Merck
Aneuploidy related transcriptional changes in endometrial cancer link low expression of chromosome 15q genes to poor survival.
Oncotarget (2017-01-01)
Karen Klepsland Mauland, Elisabeth Wik, Erling A Hoivik, Kanthida Kusonmano, Mari Kyllesø Halle, Anna Berg, Hans Kristian Haugland, Anne Margrete Øyan, Karl-Henning Kalland, Ingunn Marie Stefansson, Lars A Akslen, Camilla Krakstad, Jone Trovik, Henrica Maria Johanna Werner, Helga Birgitte Salvesen
RESUMEN
Aneuploidy is a widely studied prognostic marker in endometrial cancer (EC), however, not implemented in clinical decision-making. It lacks validation in large prospective patient cohorts adjusted for currently standard applied prognostic markers, including estrogen/progesterone receptor status (ER/PR). Also, little is known about aneuploidy-related transcriptional alterations, relevant for understanding its role in EC biology, and as therapeutic target.We included 825 EC patients with available ploidy status and comprehensive clinicopathologic characterization to analyze ploidy as a prognostic marker. For 144 patients, gene expression data were available to explore aneuploidy-related transcriptional alterations.Aneuploidy was associated with high age, FIGO stage and grade, non-endometrioid histology, ER/PR negativity, and poor survival (p-values<0.001). In patients with ER/PR negative tumors, aneuploidy independently predicted poor survival (p=0.03), lymph node metastasis (p=0.007) and recurrence (p=0.002). A prognostic 'aneuploidy signature', linked to low expression of chromosome 15q genes, was identified and validated in TCGA data.In conclusion, aneuploidy adds prognostic information in ER/PR negative EC, identifying high-risk patients that could benefit from more aggressive therapies. The 'aneuploidy signature' equally identifies these aggressive tumors and suggests a link between aneuploidy and low expression of 15q genes. Integrated analyses point at various dysregulated pathways in aneuploid EC, underlining a complex biology.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-PPP2R3A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution