Merck
HIF-driven SF3B1 induces KHK-C to enforce fructolysis and heart disease.
Nature (2015-06-18)
Peter Mirtschink, Jaya Krishnan, Fiona Grimm, Alexandre Sarre, Manuel Hörl, Melis Kayikci, Niklaus Fankhauser, Yann Christinat, Cédric Cortijo, Owen Feehan, Ana Vukolic, Samuel Sossalla, Sebastian N Stehr, Jernej Ule, Nicola Zamboni, Thierry Pedrazzini, Wilhelm Krek
RESUMEN
Fructose is a major component of dietary sugar and its overconsumption exacerbates key pathological features of metabolic syndrome. The central fructose-metabolising enzyme is ketohexokinase (KHK), which exists in two isoforms: KHK-A and KHK-C, generated through mutually exclusive alternative splicing of KHK pre-mRNAs. KHK-C displays superior affinity for fructose compared with KHK-A and is produced primarily in the liver, thus restricting fructose metabolism almost exclusively to this organ. Here we show that myocardial hypoxia actuates fructose metabolism in human and mouse models of pathological cardiac hypertrophy through hypoxia-inducible factor 1α (HIF1α) activation of SF3B1 and SF3B1-mediated splice switching of KHK-A to KHK-C. Heart-specific depletion of SF3B1 or genetic ablation of Khk, but not Khk-A alone, in mice, suppresses pathological stress-induced fructose metabolism, growth and contractile dysfunction, thus defining signalling components and molecular underpinnings of a fructose metabolism regulatory system crucial for pathological growth.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-α-actinina (sarcomérica) monoclonal antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-KHK antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution