Merck
Endoplasmic reticulum stress pathway PERK-eIF2α confers radioresistance in oropharyngeal carcinoma by activating NF-κB.
Cancer science (2017-04-19)
Qiao Qiao, Chaonan Sun, Chuyang Han, Ning Han, Miao Zhang, Guang Li
RESUMEN
Endoplasmic reticulum stress (ERS) plays an important role in the pathogenesis and development of malignant tumors, as well as in the regulation of radiochemoresistance and chemoresistance in many malignancies. ERS signaling pathway protein kinase RNA-like endoplasmic reticulum kinase (PERK)-eukaryotic initiation factor-2 (eIF2α) may induce aberrant activation of nuclear factor-κB (NF-κB). Our previous study showed that NF-κB conferred radioresistance in lymphoma cells. However, whether PERK-eIF2α regulates radioresistance in oropharyngeal carcinoma through NF-κB activation is unknown. Herein, we showed that PERK overexpression correlated with a poor prognosis for patients with oropharyngeal carcinoma (P < 0.01). Meanwhile, the percentage of the high expression level of PERK in oropharyngeal carcinoma patients resistant to radiation was higher than in patients sensitive to radiation (77.7 and 33.3%, respectively; P < 0.05). Silencing PERK and eIF2α increased the radiosensitivity in oropharyngeal carcinoma cells and increased radiation-induced apoptosis and G2/M phase arrest. PERK-eIF2α silencing also inhibited radiation-induced NF-κB phosphorylation and increased the DNA double strand break-related proteins ATM phosphorylation. NF-κB activator TNF-α and the ATM inhibitor Ku55933 offset the regulatory effect of eIF2α on the expression of radiation-induced cell apoptosis-related proteins and the G2/M phase arrest-related proteins. These data indicate that PERK regulates radioresistance in oropharyngeal carcinoma through NF-kB activation-mediated phosphorylation of eIF2α, enhancing X-ray-induced activation of DNA DSB repair, cell apoptosis inhibition and G2/M cell cycle arrest.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human EIF2AK3, AC104134.2