Merck
  • Genomic and transcriptomic analyses reveal distinct biological functions for cold shock proteins (VpaCspA and VpaCspD) in Vibrio parahaemolyticus CHN25 during low-temperature survival.
Genomic and transcriptomic analyses reveal distinct biological functions for cold shock proteins (VpaCspA and VpaCspD) in Vibrio parahaemolyticus CHN25 during low-temperature survival.
BMC genomics (2017-06-07)
Chunhua Zhu, Boyi Sun, Taigang Liu, Huajun Zheng, Wenyi Gu, Wei He, Fengjiao Sun, Yaping Wang, Meicheng Yang, Weicheng Bei, Xu Peng, Qunxin She, Lu Xie, Lanming Chen
RESUMEN
Vibrio parahaemolyticus causes serious seafood-borne gastroenteritis and death in humans. Raw seafood is often subjected to post-harvest processing and low-temperature storage. To date, very little information is available regarding the biological functions of cold shock proteins (CSPs) in the low-temperature survival of the bacterium. In this study, we determined the complete genome sequence of V. parahaemolyticus CHN25 (serotype: O5:KUT). The two main CSP-encoding genes (VpacspA and VpacspD) were deleted from the bacterial genome, and comparative transcriptomic analysis between the mutant and wild-type strains was performed to dissect the possible molecular mechanisms that underlie low-temperature adaptation by V. parahaemolyticus. The 5,443,401-bp V. parahaemolyticus CHN25 genome (45.2% G + C) consisted of two circular chromosomes and three plasmids with 4,724 predicted protein-encoding genes. One dual-gene and two single-gene deletion mutants were generated for VpacspA and VpacspD by homologous recombination. The growth of the ΔVpacspA mutant was strongly inhibited at 10 °C, whereas the VpacspD gene deletion strongly stimulated bacterial growth at this low temperature compared with the wild-type strain. The complementary phenotypes were observed in the reverse mutants (ΔVpacspA-com, and ΔVpacspD-com). The transcriptome data revealed that 12.4% of the expressed genes in V. parahaemolyticus CHN25 were significantly altered in the ΔVpacspA mutant when it was grown at 10 °C. These included genes that were involved in amino acid degradation, secretion systems, sulphur metabolism and glycerophospholipid metabolism along with ATP-binding cassette transporters. However, a low temperature elicited significant expression changes for 10.0% of the genes in the ΔVpacspD mutant, including those involved in the phosphotransferase system and in the metabolism of nitrogen and amino acids. The major metabolic pathways that were altered by the dual-gene deletion mutant (ΔVpacspAD) radically differed from those that were altered by single-gene mutants. Comparison of the transcriptome profiles further revealed numerous differentially expressed genes that were shared among the three mutants and regulators that were specifically, coordinately or antagonistically modulated by VpaCspA and VpaCspD. Our data also revealed several possible molecular coping strategies for low-temperature adaptation by the bacterium. This study is the first to describe the complete genome sequence of V. parahaemolyticus (serotype: O5:KUT). The gene deletions, complementary insertions, and comparative transcriptomics demonstrate that VpaCspA is a primary CSP in the bacterium, while VpaCspD functions as a growth inhibitor at 10 °C. These results have improved our understanding of the genetic basis for low-temperature survival by the most common seafood-borne pathogen worldwide.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,3-Diaminopropane, ≥99%