Merck
  • Simultaneous knock-down of six β-galactosidase genes in petunia petals prevents loss of pectic galactan but decreases petal strength.
Simultaneous knock-down of six β-galactosidase genes in petunia petals prevents loss of pectic galactan but decreases petal strength.
Plant physiology and biochemistry : PPB (2017-03-04)
Erin M O'Donoghue, Sheryl D Somerfield, Simon C Deroles, Paul W Sutherland, Ian C Hallett, Zoë A Erridge, David A Brummell, Donald A Hunter
RESUMEN
Galactose (Gal) is incorporated into cell wall polysaccharides as flowers open, but then is lost because of β-galactosidase activity as flowers mature and wilt. The significance of this for flower physiology resides in the role of galactan-containing polysaccharides in the cell wall, which is still largely unresolved. To investigate this, transcript accumulation of six cell wall-associated β-galactosidases was simultaneously knocked down in 'Mitchell' petunia (Petunia axillaris x (P. axillaris x P. hybrida)) flower petals. The multi-PhBGAL RNAi construct targeted three bud- and three senescence-associated β-galactosidase genes. The petals of the most down-regulated line (GA19) were significantly disrupted in galactose turnover during flower opening, and at the onset of senescence had retained 86% of their galactose compared with 20% in the controls. The Gal content of Na
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Chlorophenol Red, indicator grade