Merck
  • TRIM22 confers poor prognosis and promotes epithelial-mesenchymal transition through regulation of AKT/GSK3β/β-catenin signaling in non-small cell lung cancer.
TRIM22 confers poor prognosis and promotes epithelial-mesenchymal transition through regulation of AKT/GSK3β/β-catenin signaling in non-small cell lung cancer.
Oncotarget (2017-10-06)
Li Liu, Xiao-Ming Zhou, Fang-Fei Yang, Yuan Miao, Yan Yin, Xue-Jun Hu, Gang Hou, Qiu-Yue Wang, Jian Kang
RESUMEN
Expression pattern and biological roles of TRIM22 remains unknown in most human cancers. The present study aims to discover its clinical significance and function in human non-small cell lung cancer (NSCLC). Immunohistochemistry was used to examine TRIM22 expression in 126 cases of NSCLC specimens. TRIM22 protein was upregulated in 70/126 (55.6%) non-small cell lung cancer tissues compared with normal lung tissue. TRIM22 overexpression was associated with advanced TNM stage, positive nodal metastasis and poor prognosis. Plasmid and siRNA transfection were performed in lung cancer cell lines. TRIM22 overexpression promoted proliferation, colony formation and invasion in A549 cells. While its depletion exhibited the opposite effects in H1299 cell line. TRIM22 overexpression promoted cell cycle progression through regulation of cyclin D1, cyclin E and p27. TRIM22 also changed the expression of epithelial to mesenchymal transition (EMT) markers including E-cadherin N-cadherin, Vimentin and Snail. Furthermore, TRIM22 activated PI3K/AKT/GSK3β/β-catenin oncogenic signaling pathways. Treatment with PI3K inhibitor LY294002 and β-catenin siRNA blocked the effects of TRIM22 on EMT in TRIM22-overexpressing cells. In conclusion,TRIM22 serves as an important oncoprotein and a promoter of cell proliferation and invasion through AKT/ GSK3β/β-catenin induced EMT in NSCLC.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human TRIM22