Merck
  • Structural Analysis of Variable Domain Glycosylation of Anti-Citrullinated Protein Antibodies in Rheumatoid Arthritis Reveals the Presence of Highly Sialylated Glycans.
Structural Analysis of Variable Domain Glycosylation of Anti-Citrullinated Protein Antibodies in Rheumatoid Arthritis Reveals the Presence of Highly Sialylated Glycans.
Molecular & cellular proteomics : MCP (2016-12-14)
Lise Hafkenscheid, Albert Bondt, Hans U Scherer, Tom W J Huizinga, Manfred Wuhrer, René E M Toes, Yoann Rombouts
RESUMEN
Recently, we showed the unexpectedly high abundance of N-linked glycans on the Fab-domain of Anti-Citrullinated Protein Antibodies (ACPA). As N-linked glycans can mediate a variety of biological functions, we now aimed at investigating the structural composition of the Fab-glycans of ACPA-IgG to better understand their mediated biological effects. ACPA-IgG and noncitrulline specific (control) IgG from plasma and/or synovial fluid of nine ACPA positive rheumatoid arthritis patients were affinity purified. The N-linked glycosylation of total, Fc and F(ab')
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2-Hydroxy-5-methoxybenzoic acid, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad