Merck
Large-scale production of recombinant human lactoferrin from high-expression, marker-free transgenic cloned cows.
Scientific reports (2017-09-08)
Ming Wang, Zhaolin Sun, Tian Yu, Fangrong Ding, Ling Li, Xi Wang, Mingbo Fu, Haiping Wang, Jinming Huang, Ning Li, Yunping Dai
RESUMEN
Human lactoferrin (hLF) is a valuable protein for pharmaceutical products and functional foods, and worldwide demand for this protein has steadily increased. However, large-scale recombinant human lactoferrin (rhLF) production using current animal bioreactor techniques is limited by the low expression of foreign proteins, the use of antibiotic resistance genes and the down-regulation of endogenous milk proteins. Here, we generated a herd of marker-free, hLF bacterial artificial chromosome (BAC) transgenic cloned cows, as confirmed by Polymerase chain reaction, Southern blot and Western blot analyses. These transgenic cloned cows produced rhLF in milk at concentrations of 4.5-13.6 g/L. Moreover, the total protein content of the milk was increased. Over two hundred transgenic cloned cows were propagated by multiple ovulation and embryo transfer (MOET). A total of 400-450 g of rhLF protein, which shows similar enzymatic activity to natural hLF in iron binding and release, can be purified on a large scale from >100 L of milk per day. Our results suggested that transgenic bovine mammary bioreactors have the potential for large-scale protein production.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Human LTF / Lactotransferrin ELISA Kit