Merck
Beneficial effects of natural eggshell membrane (NEM) on multiple indices of arthritis in collagen-induced arthritic rats.
Modern rheumatology (2016-11-17)
Karen J Wedekind, Kevin J Ruff, Cindy A Atwell, Joseph L Evans, Alison M Bendele
RESUMEN
This study was performed to evaluate the potential efficacy of natural eggshell membrane (NEM) in collagen-induced arthritic rats, a well-established rodent model of inflammation and rheumatoid arthritis. Rats with developing type II collagen-induced arthritis (CIA) were treated once daily by oral gavage on study days -14 to 17 with vehicle or NEM (52 mg/kg body weight). Rats were euthanized on study day 17. Efficacy was assessed by daily ankle caliper measurements, ankle diameter expressed as area under the curve (AUC Treatment with NEM resulted in significant beneficial effects on the daily ankle diameter measurements and ankle diameter AUC. Ankle and knee histopathology scores were significantly reduced (36% and 43% reduction of summed individual histopathology scores for ankle and knee, respectively; p < 0.05) toward normal for rats given NEM compared to vehicle controls. The percent reduction of serum CTXII, COMP, and A2M in NEM-treated rats ranged from 30% to 72% (p < 0.05). NEM significantly improved multiple aspects of inflammatory arthritis including inflammation, pannus, cartilage damage, bone resorption, and periosteal bone formation. This study provides further support for the use of CTXII, COMP, and A2M as relevant biomarkers that were responsive to NEM.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Toluidine Blue, suitable for microscopy, 8.74% (ZN (THEORY))
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP