Merck
Targeting the cancer stem cell marker, aldehyde dehydrogenase 1, to circumvent cisplatin resistance in NSCLC.
Oncotarget (2017-10-27)
Lauren MacDonagh, Michael F Gallagher, Brendan Ffrench, Claudia Gasch, Eamon Breen, Steven G Gray, Siobhan Nicholson, Niamh Leonard, Ronan Ryan, Vincent Young, John J O'Leary, Sinead Cuffe, Stephen P Finn, Kenneth J O'Byrne, Martin P Barr
RESUMEN
Non-small cell lung cancer (NSCLC) accounts for a large proportion of cancer deaths and is characterized by low treatment response rates and poor overall prognosis. In the absence of specific treatable mutations, cisplatin-based chemotherapy plays an important role in the treatment of this disease. Unfortunately, the development of resistance has become a major therapeutic challenge in the use of this cytotoxic drug. Elucidating the mechanisms underlying this resistance phenotype, may result in the development of novel agents that enhance sensitivity to cisplatin in lung cancer patients. In this study, targeting the cancer stem cell activity of aldehyde dehydrogenase 1 (ALDH1) was investigated as a strategy to overcome chemoresistance in NSCLC. Tumors from NSCLC patients showed an increase in their profile of pluripotent stemness genes. Cisplatin exposure induced the emergence or expansion of an ALDH1-positive subpopulation in cisplatin sensitive and resistant NSCLC cell lines, respectively, further enhancing cisplatin resistance. Using the Aldefluor assay and FACS analysis, ALDH1 subpopulations were isolated and evaluated in terms of stem cell characteristics. Only ALDH1-positive cells exhibited asymmetric division, cisplatin resistance and increased expression of stem cell factors
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
cis-11-Methyl-2-dodecenoic acid, ≥90.0% (HPLC)