Merck
  • Reduced production and uptake of lactate are essential for the ability of WNT5A signaling to inhibit breast cancer cell migration and invasion.
Reduced production and uptake of lactate are essential for the ability of WNT5A signaling to inhibit breast cancer cell migration and invasion.
Oncotarget (2017-10-27)
Chandra Prakash Prasad, Katja Södergren, Tommy Andersson
RESUMEN
Here we investigated the impact of WNT5A signaling on aerobic glycolysis and evaluated its effects on breast cancer cell migration/invasion. WNT5A signaling reduced migration and lactate production and caused selective down-regulation of the glycolytic enzyme phosphofructokinase platelet-type (PFKP). These events occurred in parallel with a WNT5A-induced inhibition of β-catenin signaling. Support for essential involvement of β-catenin and PFKP in lactate production and migration/invasion was obtained by siRNA knockdown of their expression. To also explore the effect of non-tumor cell-derived lactate, we added exogenous lactate to the cells and noted an increase in migration that was significantly impaired by recombinant WNT5A in parallel with a down-regulation of the lactate transporter monocarboxylate transporter 1 (MCT1). Interestingly enough, the drug-candidate Foxy5 (WNT5A-mimic hexapeptide) also inhibited breast cancer cell migration in the presence of exogenous lactate, suggesting a therapeutic potential for Foxy5 in managing breast tumors with high glycolytic activity. Overall, we demonstrated that WNT5A signaling (via a β-catenin-PFKP axis) reduces lactate production and lowers the expression of MCT1, a carrier mediating the uptake of lactate from the tumor microenvironment. These effects of WNT5A are essential for its ability to impair breast cancer migration/invasion even in an environment with elevated lactate levels.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PFKP