Merck
Glucagon-like peptide receptor agonists attenuate advanced glycation end products-induced inflammation in rat mesangial cells.
BMC pharmacology & toxicology (2017-10-27)
Jui-Ting Chang, Yao-Jen Liang, Chia-Yu Hsu, Chao-Yi Chen, Po-Jung Chen, Yi-Feng Yang, Yen-Lin Chen, Dee Pei, Jin-Biou Chang, Jyh-Gang Leu
RESUMEN
Hyperglycemia-induced advanced glycation end products (AGEs) and receptor for AGEs (RAGE) production play major roles in progression of diabetic nephropathy. Anti-RAGE effect of peroxisome proliferator-activated receptor-delta (PPARδ) agonists was shown in previous studies. PPARδ agonists also stimulate glucagon-like peptide-1 (GLP-1) secretion from human intestinal cells. In this study, the individual and synergic anti-inflammatory effects of GLP-1 receptor (exendin-4) and PPARδ (L-165,041) agonists in AGE-treated rat mesangial cells (RMC) were investigated. The results showed both exendin-4 and L-165,041 significantly attenuated AGE-induced IL-6 and TNF-α production, RAGE expression, and cell death in RMC. Similar anti-inflammatory potency was seen between 0.3 nM exendin-4 and 1 μM L-165,041. Synergic effect of exendin-4 and L-165,041 was shown in inhibiting cytokines production, but not in inhibiting RAGE expression or cell death. These results suggest that both GLP-1 receptor and PPARδ agonists have anti-inflammatory effect on AGE-treated rat mesangial cells.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-165,041, ≥98% (HPLC), powder