Merck
Tissue-type plasminogen activator modulates macrophage M2 to M1 phenotypic change through annexin A2-mediated NF-κB pathway.
Oncotarget (2017-11-21)
Ling Lin, Kebin Hu
RESUMEN
Macrophage accumulation is one of the hallmarks of progressive kidney disease. In response to injury, macrophages undergo a phenotypic polarization to become two functionally distinct subsets: M1 and M2 macrophages. Macrophage polarization is a dynamic process, and recent work indicates that macrophages, in response to kidney injury, can shift their polarity. However, the underlying mechanisms remain largely unknown. Tissue-type plasminogen activator (tPA), a protease up-regulated in the chronically injured kidneys, has been shown to preferably promote M1 macrophage accumulation and renal inflammation. We hypothesized that tPA may be an endogenous factor that modulates macrophage M2 to M1 phenotypic change contributing to the accumulation of M1 macrophages in the injured kidneys. It was found that obstruction-induced renal M1 chemokine expression was alleviated in tPA knockout mice, and these knockout mice displayed increased M2 markers.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human LRP1