Merck
Size-Dependent Biexciton Quantum Yields and Carrier Dynamics of Quasi-Two-Dimensional Core/Shell Nanoplatelets.
ACS nano (2017-08-09)
Xuedan Ma, Benjamin T Diroll, Wooje Cho, Igor Fedin, Richard D Schaller, Dmitri V Talapin, Stephen K Gray, Gary P Wiederrecht, David J Gosztola
RESUMEN
Quasi-two-dimensional nanoplatelets (NPLs) possess fundamentally different excitonic properties from zero-dimensional quantum dots. We study lateral size-dependent photon emission statistics and carrier dynamics of individual NPLs using second-order photon correlation (g
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Oleilamina, technical grade, 70%
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Ácido oleico, technical grade, 90%
Sigma-Aldrich
Sodium myristate, ≥99%