Merck
Binding of A 1,4-dihydropyridine calcium channel activator, (-) S Bay K 8644, to cardiac preparations.
Biochemical and biophysical research communications (1989-01-16)
J Ferrante, E Luchowski, A Rutledge, D J Triggle
RESUMEN
The 1,4-dihydropyridine Ca2+ channel activator, (-) [3H]Bay K 8644, binds to cardiac membranes and polarized [5 mM K+] and depolarized [50 mM K+] cardiac cells. Binding to microsomal membranes at 25 degrees C indicates a single set of binding sites, KD = 2.9 x 10(-9) M and a site density, 337 fmoles/mg protein, not different from that measured by antagonist 1,4-dihydropyridines. Binding to neonatal rat myocytes at 37 degrees C was independent of membrane potential with a KD value of 5 x 10(-8)M and a site density, 63 fmoles/mg protein, not significantly different from that measured by PN 200 110. These results indicate that 1,4-dihydropyridine activators and antagonists label the same number of binding sites in cardiac tissue, but that activator binding to intact myocytes is voltage-independent.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(S)-(−)-Bay K8644, ≥98% (HPLC), solid