Merck
Rebamipide protects against glaucoma eyedrop-induced ocular surface disorders in rabbits.
PloS one (2017-10-20)
Ichiro Kawaguchi, Akira Kobayashi, Tomomi Higashide, Yasuhiro Takeji, Kazushi Sakurai, Chiaki Kawaguchi, Kazuhisa Sugiyama
RESUMEN
This study aimed to determine if rebamipide eyedrops can improve ocular surface damage caused by the use of glaucoma eyedrops. Female Kbl:Dutch rabbits were used to evaluate glaucoma eyedrop-induced ocular surface damage; one eye of each rabbit was untreated and the other was administered glaucoma eyedrops for 30 days. To evaluate the effects of rebamipide on ocular surface damage, one eye of each rabbit was administered vehicle-treated glaucoma eyedrops and the other was administered rebamipide-treated glaucoma eyedrops for 30 days. Corneal and conjunctival epithelial damage was evaluated using fluorescein and rose bengal staining, respectively. Conjunctival inflammation was observed by light microscopy with hematoxylin-eosin staining. Dark cells (in which the corneal microvilli were damaged) were analyzed by scanning electron microscopy. There were no significant differences in fluorescein staining between the untreated and glaucoma eyedrop-treated groups; however, rose bengal staining and the number of inflammatory cells in the conjunctiva significantly increased after glaucoma eyedrop treatment. There was a four-fold increase in the number of dark cells in the glaucoma eyedrop-treated group compared to untreated. In contrast, in the conjunctiva of the rebamipide-treated glaucoma eyedrop group, rose bengal staining scores, the number of inflammatory cells, and the number of dark cells were decreased compared to the vehicle-treated glaucoma eyedrop group. Results from our in vivo rabbit study demonstrated that short-term use of glaucoma eyedrops induces corneal epithelium disorders at the cellular level, but that simultaneous use of rebamipide has the potential to protect and repair the ocular surface.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dorzolamide, ≥98% (HPLC)