Merck
ACK1/TNK2 Regulates Histone H4 Tyr88-phosphorylation and AR Gene Expression in Castration-Resistant Prostate Cancer.
Cancer cell (2017-06-14)
Kiran Mahajan, Pavani Malla, Harshani R Lawrence, Zhihua Chen, Chandan Kumar-Sinha, Rohit Malik, Sudhanshu Shukla, Jongphil Kim, Domenico Coppola, Nicholas J Lawrence, Nupam P Mahajan
RESUMEN
The androgen receptor (AR) is critical for the progression of prostate cancer to a castration-resistant (CRPC) state. AR antagonists are ineffective due to their inability to repress the expression of AR or its splice variant, AR-V7. Here, we report that the tyrosine kinase ACK1 (TNK2) phosphorylates histone H4 at tyrosine 88 upstream of the AR transcription start site. The WDR5/MLL2 complex reads the H4-Y88-phosphorylation marks and deposits the transcriptionally activating H3K4-trimethyl marks promoting AR transcription. Reversal of the pY88-H4 epigenetic marks by the ACK1 inhibitor (R)-9bMS-sensitized naive and enzalutamide-resistant prostate cancer cells and reduced AR and AR-V7 levels to mitigate CRPC tumor growth. Thus, a feedforward ACK1/pY88-H4/WDR5/MLL2/AR epigenetic circuit drives CRPC and is necessary for maintenance of the malignant state.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
(R)-9bMS, ≥98% (HPLC)