Merck
Increase in complement iC3b is associated with anti-inflammatory cytokine expression during late pregnancy in mice.
PloS one (2017-05-26)
Keigo Nakamura, Kazuya Kusama, Rulan Bai, Sadamasa Ishikawa, Sayuri Fukushima, Yoshihito Suda, Kazuhiko Imakawa
RESUMEN
Immunological tolerance between fetal allograft and mother is crucial for pregnancy establishment and maintenance; however, these mechanisms particularly those during the latter part of pregnancy have not been definitively elucidated. The aim of this study was to examine the presence and potential function of innate immunity characteristic to the middle to late pregnancy. We first characterized up-regulated proteins in decidua from day 11 pregnant (P11) mice using 2D-PAGE, followed by MALDI-TOF/MS analysis. These analyses identified increased complement component 3 (C3) and its derivatives in P11 decidua. We then found that in the decidual tissues, C3 mRNA increased on P15 and remained high on P19. C3 is converted to C3b and then iC3b by complement component factor I (Cfi) and complement receptor 1-like protein (Crry), both of which were present in P19 placentas. In addition, iC3b proteins and its receptor CR3 (Cd11b/Cd18) in decidual and placental tissues increased toward the latter phase of pregnancy. Moreover, CR3 subunit CD11b protein was predominantly localized to spongiotrophoblast layer in the P19 placenta. Because iC3b is known to induce anti-inflammatory cytokine production, the analysis was extended to examine changes in pro- and anti-inflammatory cytokines, Il12, Il10, and Tgfb1. Il12 expression decreased in P15 and P19 placenta, while high mRNA expression of Il10 and Tgfb1 was found in P19 placental tissues. Furthermore, placental Il10 and Tgfb1 mRNAs were down-regulated when pregnant mice were treated with an anti-C3 antibody, detecting C3, C3b and iC3b. These results indicated that C3 derivatives, in particular, iC3b and its receptor CR3 were up-regulated at the fetal-maternal interface, and suggest that iC3b may regulate the placental expression of anti-inflammatory cytokines, IL10 and TGFB1, during the latter phase of pregnancy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution