Merck
[Study on volatile components of Lonicerae Japonicae Flos in bud stage extended type].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2017-09-22)
Jing-Ru Zhang, Gen-Ben Bai, Wei-Dong Li, Zhong-Shuang Zhang, Jin-Rui Gu, Cai-Qiao Zhang
RESUMEN
Volatile components of Lonicerae Japonicae Flos in bud stage extended type Beihua 1 were determined by the headspace solid-phase micro-extraction, compared with traditional cultivar Damaohua. There are fifty-two volatile compounds were identified and the relative content of the volatiles was calculated by the area normalization method. Thirty-nine compounds were found in Beihua 1, whereas thirty-three components in Damaohua. Total twenty identical compounds existed in Beihua 1 and Damaohua. The contents of alcohols and hydrocarbons of Beihua 1 were higher significantly than that of Damaohua, while significantly lower than that of Damaohua in ketones content. Besides, twenty components were only detected in Beihua 1, such as methyl nicotinate, hexadecanoic acid, methyl ester,acetophenone, nonanoic acid.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
Montaje de fibra SPME, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME fiber assembly polyacrylate (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME fiber assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME fiber assembly polyacrylate (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
Kit de surtido de fibra SPME StableFlex, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
Fibra de carboxeno/polidimetilsiloxano (DVB/CAR/PDMS) de SPME, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
Filtro SPME Polidimetilsiloxano (PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
Kit de surtido de fibra SPME StableFlex, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assortment kit 4, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub