Product Name
Eppendorf Platform for sticky pads, for use with Innova® S44i
material
metal
feature
to hold flasks
for use with Innova® S44i
manufacturer/tradename
Eppendorf® S44I040009
packaging
pack of -1 ea
Application
Platform to hold flasks / plates on shaker
General description
Shaker platform with surface sutable for use with screwed-on accessories such as racks and flask clamps or with sticky pads, re-usable siliconized sheets that will last several years. The Sticky Pad can be easily removed and re-positioned simply by peeling it from the surface.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc.
Choose from one of the most recent versions:
Already Own This Product?