Skip to Content
Merck

LEYH9B0CD427

Leyan / Shanghai Haohong Scientific Co.,Ltd

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methyldec-9-enoic acid

Sign In to View Organizational & Contract Pricing

Select a Size

Product LEYH9B0CD427 is not currently sold in your country.

Product LEYH9B0CD427 is not currently sold in your country.

Iupac Name

(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-2-methyl-9-decenoic acid

Inchi Code

1S/C26H31NO4/c1-3-4-5-6-7-12-17-26(2,24(28)29)27-25(30)31-18-23-21-15-10-8-13-19(21)20-14-9-11-16-22(20)23/h3,8-11,13-16,23H,1,4-7,12,17-18H2,2H3,(H,27,30)(H,28,29)/t26-/m0/s1

InChI key

MADFVGMQNXRFAF-SANMLTNESA-N

storage temp.

2-8C, stored under nitrogen

Purity

98%

Country of Origin

CN

Shipping Temperature

2-8C

Physical Form

Liquid

Product Link

https://www.leyan.com/288617-75-4.html

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.leyan.com/288617-75-4.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library