Skip to Content
Merck

14-680

HIPK3 Protein, active, 10 µg

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target HIPK3

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... HIPK3(10114)

General description

49.7kDa
FUNCTION: SwissProt: Q9H422 # Seems to negatively regulate apoptosis by promoting FADD phosphorylation. Enhances androgen receptor-mediated transcription. May act as a transcriptional corepressor for NK homeodomain transcription factors.
SIZE: 1215 amino acids; 133743 Da
SUBUNIT: Interacts with Nkx1-2. Interacts with FAS and DAXX. Probably part of a complex consisting of HIPK3, FAS and FADD. Interacts with and stabilizes ligand-bound androgen receptor (AR) (By similarity). Interacts with UBL1/SUMO-1.
SUBCELLULAR LOCATION: Cytoplasm. Nucleus.
TISSUE SPECIFICITY: Overexpressed in multidrug resistant cells. Highly expressed in heart and skeletal muscle, and at lower levels in placenta, pancreas, brain, spleen, prostate, thymus, testis, small intestine, colon and leukocytes. Not found in liver and lung.
PTM: Autophosphorylated, but autophosphorylation is not required for catalytic activity (By similarity). & May be sumoylated (By similarity).
SIMILARITY: SwissProt: Q9H422 ## Belongs to the protein kinase superfamily. CMGC Ser/Thr protein kinase family. HIPK subfamily. & Contains 1 protein kinase domain.

Preparation Note

Stable 6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.
routinely evaluated by phosphorylation of MBP (Catalog # 13-104)

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service