Skip to Content
Merck

77558

Pipecolinic acid

new

analytical standard

Synonym(s):

(±)-Piperidine-2-carboxylic acid, 2-Piperidinecarboxylic acid, Homoproline, Pipecolic acid free base

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
Beilstein:
81095
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
LC/MS: suitable
liquid chromatography (LC): suitable
mass spectrometry (MS): suitable

mp

282 °C (dec.) (lit.)

application(s)

clinical
food and beverages

format

neat

SMILES string

OC(=O)C1CCCCN1

InChI

1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)

InChI key

HXEACLLIILLPRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pipecolic acid (piperidine-2-carboxylic acid) is a non-proteinogenic cyclic amino acid formed during lysine catabolism and further processed in peroxisomes. Elevated levels are a useful biomarker for peroxisomal biogenesis disorders and can also appear in pyridoxine-dependent epilepsy and certain liver-related conditions. Testing pipecolic acid in serum or urine—typically by LC–MS/MS—supports differential diagnosis and monitoring.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service