Skip to Content
Merck

BDPH9BCD3C07

Bide Pharmatech

4-Methoxybenzo[d]isothiazol-3-amine

Synonym(s):

4-methoxybenzo[d]isothiazol-3(2H)-imine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O1N2S1
CAS Number:
Molecular Weight:
180.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H8N2OS/c1-11-5-3-2-4-6-7(5)8(9)10-12-6/h2-4H,1H3,(H2,9,10)

Inchi Key

ZMIMCZXQNAIZQB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library