Skip to Content
Merck

BDPH9BCD7DFC

Bide Pharmatech

6-(Difluoromethoxy)pyridin-2-amine

Synonym(s):

6-(difluoromethoxy)pyridin-2-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N2F2
CAS Number:
Molecular Weight:
160.12
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6F2N2O/c7-6(8)11-5-3-1-2-4(9)10-5/h1-3,6H,(H2,9,10)

Inchi Key

QMOFLAKXMLKVTC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Colorless to Pale-yellow to Yellow-brown Liquid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 6-(Difluoromethoxy)pyridin-2-amine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library