Skip to Content
Merck

BDPH9BCDBE8B

Bide Pharmatech

4-Nitrophthalonitrile

Synonym(s):

4-nitrophthalonitrile

Select a Size

Change View

About This Item

Linear Formula:
C8H3O2N3
CAS Number:
Molecular Weight:
173.13
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H3N3O2/c9-4-6-1-2-8(11(12)13)3-7(6)5-10/h1-3H

Inchi Key

NTZMSBAAHBICLE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder



Still not finding the right product?

Explore all of our products under 4-Nitrophthalonitrile




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library