Skip to Content
Products
Cart
0
FR
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Bromide bromate solution
Products
Product Category
reagents
Brand
Supelco
Physical Form
Product Line
Quality Segment
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.09905
Bromide bromate solution,
for 1000 ml, c(Br
2
) = 0,05 mol/l (0.1 N) Titrisol
®
Expand
View Pricing
An unknown error has occured.