Skip to Content
Merck
Potassium bromate solution 1/60 M KBrO3 (0.1 N), Titripur®, reag. USP

Potassium bromate solution

Potassium bromate solution 1/60 M KBrO3 (0.1 N), Titripur®, reag. USP

Products

Brand
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing