Merck
  • Spectroscopic and Molecular Docking Investigation on the Noncovalent Interaction of Lysozyme with Saffron Constituent "Safranal".
Spectroscopic and Molecular Docking Investigation on the Noncovalent Interaction of Lysozyme with Saffron Constituent "Safranal".
ACS omega (2020-05-05)
Mohd Sajid Ali, Hamad A Al-Lohedan
ABSTRACT
Owing to the various beneficial properties of the popular spice saffron, the interaction of safranal, a secondary metabolite of the former, with hen egg white lysozyme was investigated. The formation of a complex was evidenced by UV-visible spectroscopy. Fluorescence quenching experiments were also performed to understand the binding mechanism and to evaluate the forces involved in binding. The strong absorption of safranal in the range of excitation and emission wavelengths of lysozyme fluorescence required the correction of the inner filter effect for fluorescence spectra to obtain the apparent extent of binding. There was a considerable difference between the observed spectra and corrected spectra, and a similar observation was found in the case of synchronous fluorescence spectra. From the analysis of quenching data, it was found that the mechanism involved in quenching was static with 1:1 binding between them. The interaction was found to be driven, mainly, by hydrophobic forces and hydrogen bonding. Safranal had negligible impact on the secondary structure of lysozyme. The interaction was also studied by molecular docking, and the results were in good agreement with the results obtained experimentally. The binding site of safranal was in the big hydrophobic cavity of lysozyme. The amino acids involved in the interaction were Asp52, Ile58, Gln57, Asn59, Trp62, Trp63, Trp108, Ile98, Asp101, and Ala107.
Sigma-Aldrich
Lysozyme from chicken egg white, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Safranal, ≥90%, stabilized