Merck
  • Synthesis and antifungal activity of some substituted phenothiazines and related compounds.
Synthesis and antifungal activity of some substituted phenothiazines and related compounds.
European journal of medicinal chemistry (2010-11-26)
Gabriela P Sarmiento, Roxana G Vitale, Javier Afeltra, Graciela Y Moltrasio, Albertina G Moglioni
ABSTRACT
Several phenothiazines and related compounds were synthesized and their antifungal activity was evaluated in vitro. The results observed for α-chloro-N-acetyl phenothiazine led us to choose this compound as a lead in the search of antifungal agents.
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog