Merck
  • A novel fibrin(ogen)olytic trypsin-like protease from Chinese oak silkworm (Antheraea pernyi): purification and characterization.
A novel fibrin(ogen)olytic trypsin-like protease from Chinese oak silkworm (Antheraea pernyi): purification and characterization.
Biochemical and biophysical research communications (2014-02-05)
Peng Geng, Lan Lin, Yuan Li, Qi Fan, Naihong Wang, Lixin Song, Wenli Li
ABSTRACT
A novel fibrin(ogen)olytic protease from Antheraea pernyi (important economically insect), named cocoonase, was isolated by a combination of ion-exchange chromatography and gel filtration. Furthermore, the characterization of cocoonase was investigated using fibrin(ogen)olytic, thrombolysis, and hemorrhagic assays. The NH2-terminal sequence (IVGGY SVTID KAPYQ) was established by Edman degradation. Based on the N-terminal sequencing, cocoonase cDNA has been cloned by means of RT-PCR and 5'RACE. It is composed of 261 amino acid residues and possesses the structural features of trypsin-like serine protease. The purified cocoonase showed specific esterase activity on N-β-benzoyl-l-arginine ethyl (BAEE), and the kinetic constants, Km and Vmax were 2.577 × 10(-3)mol/L and 4.09 × 10(-3)μmol/L/s, respectively. Cocoonase showed strong activities on both fibrin and fibrinogen, preferentially hydrolyzed Aα and Bβ chains followed by γ-chains of fibrinogen. Cocoonase exhibited a thrombolysis activity both in vitro (blood-clot lysis activity assay) and in vivo (carrageenan-induced thrombosis model). These findings indicate that A. pernyi cocoonase ia a novel fibrin(ogen)olytic enzyme and may have a potential clinical application as an antithrombotic agent.
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
Sigma-Aldrich
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin from porcine pancreas, tablet, 1 mg tablet
Sigma-Aldrich
TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin from porcine pancreas, ~1500 U/mg
Sigma-Aldrich
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder