Merck
  • Adenoviral vectors coated with cationic PEG derivatives for intravaginal vaccination against HIV-1.
Adenoviral vectors coated with cationic PEG derivatives for intravaginal vaccination against HIV-1.
Biomaterials (2014-06-16)
Zhaolu Xie, Zhonghua Ji, Zhirong Zhang, Tao Gong, Xun Sun
ABSTRACT
Mucus layer coating the vaginal epithelium represents a barrier for intravaginally delivered recombined adenoviral (rAd) vectors, but it could be overcome by proper polyethylene glycol (PEG) modification. Here we synthesized two cationic PEG derivatives, amino-(EO)n/(AGE)m-Cyss (APCs). The polymers contained neutral linear PEG (2-5 kDa) to provide a hydrophilic surface and amine pendants to provide positive charge for coating negatively charged rAd by physical adsorption. Given proper molecular composition, the polymer (5k-APC) could coat rAd without causing aggregation, facilitating its mucus penetrating ability and enhancing gene expression both in vitro and in vivo. With HIVgag as the model antigen, the polymer-rAd complexes were administered intravaginally to elicit both systemic and mucosal immune responses. 5k-APC-rAd immunization elicited robust HIVgag-specific cellular responses and also induced higher antigen-specific serum IgG. More importantly, mice immunized with 5k-APC-rAd showed higher level of IgA in vaginal lavage fluid. These findings suggest that 5k-APC-rAd is a promising system for intravaginal immunization against infectious diseases such as HIV within the vaginal tract.
Sigma-Aldrich
Cysteamine hydrochloride, ≥98% (titration)
Sigma-Aldrich
Cysteamine hydrochloride, ≥97.0% (RT)
Sigma-Aldrich
Cysteamine hydrochloride, BioXtra
Medroxyprogesterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
Medroxyprogesterone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Medroxyprogesterone acetate, United States Pharmacopeia (USP) Reference Standard
Medroxyprogesterone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Human IFN γ ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IFN γ ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
Medroxyprogesterone 17-acetate, VETRANAL®, analytical standard
Sigma-Aldrich
Medroxyprogesterone 17-acetate, ≥97% (HPLC)
Sigma-Aldrich
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit, for serum, plasma, cell culture supernatants and urine