Merck
  • Enhancing the efficacy of electrolytic chlorination for ballast water treatment by adding carbon dioxide.
Enhancing the efficacy of electrolytic chlorination for ballast water treatment by adding carbon dioxide.
Marine pollution bulletin (2015-04-07)
Hyung-Gon Cha, Min-Ho Seo, Heon-Young Lee, Ji-Hyun Lee, Dong-Sup Lee, Kyoungsoon Shin, Keun-Hyung Choi
ABSTRACT
We examined the synergistic effects of CO2 injection on electro-chlorination in disinfection of plankton and bacteria in simulated ballast water. Chlorination was performed at dosages of 4 and 6ppm with and without CO2 injection on electro-chlorination. Testing was performed in both seawater and brackish water quality as defined by IMO G8 guidelines. CO2 injection notably decreased from the control the number of Artemia franciscana, a brine shrimp, surviving during a 5-day post-treatment incubation (1.8 and 2.3 log10 reduction in seawater and brackish water, respectively at 6ppm TRO+CO2) compared with water electro-chlorinated only (1.2 and 1.3 log10 reduction in seawater and brackish water, respectively at 6ppm TRO). The phytoplankton Tetraselmis suecica, was completely disinfected with no live cell found at >4ppm TRO with and without CO2 addition. The effects of CO2 addition on heterotrophic bacterial growth was not different from electro-chlorination only. Total residual oxidant concentration (TRO) more rapidly declined in electro-chlorination of both marine and brackish waters compared to chlorine+CO2 treated waters, with significantly higher amount of TRO being left in waters treated with the CO2 addition. Total concentration of trihalomethanes (THMs) and haloacetic acids (HAAs) measured at day 0 in brackish water test were found to be 2- to 3-fold higher in 6ppm TRO+CO2-treated water than in 6ppm TRO treated water. The addition of CO2 to electro-chlorination may improve the efficiency of this sterilizing treatment of ballast water, yet the increased production of some disinfection byproducts needs further study.
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain