Merck
Vinylation of aryl bromides using an inexpensive vinylpolysiloxane.
Organic letters (2005-12-31)
Scott E Denmark, Christopher R Butler
ABSTRACT
[reaction: see text] A mild and general method for the palladium-catalyzed vinylation of aryl bromides has been developed. The use of tetrabutylammonium fluoride (TBAF) as the activator and an inexpensive and nontoxic vinyl donor, 1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane (D(4)(V), 1), allows for a general and high-yielding preparation of substituted styrenes.
Sigma-Aldrich
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
Pricing and availability is not currently available.