Merck
  • Prediction of the metabolic clearance of benzophenone-2, and its interaction with isoeugenol and coumarin using cryopreserved human hepatocytes in primary culture.
Prediction of the metabolic clearance of benzophenone-2, and its interaction with isoeugenol and coumarin using cryopreserved human hepatocytes in primary culture.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2016-02-02)
Georges de Sousa, Sophie Teng, Romain Salle-Siri, Alexandre Pery, Roger Rahmani
ABSTRACT
Benzophenone-2 (BP2) is widely used as a UV screen in both industrial products and cosmetic formulations, where it is frequently found associated with fragrance compounds, such as isoeugenol and coumarin. BP2 is now recognized as an endocrine disruptor, but to date, no information has been reported on its fate in humans. The intrinsic clearance (Clint) and metabolic interactions of BP2 were explored using cryopreserved human hepatocytes in primary cultures. In vitro kinetic experiments were performed to estimate the Michaelis-Menten parameters. The substrate depletion method demonstrated that isoeugenol was cleared more rapidly than BP2 or coumarin (Clint = 259, 94.7 and 0.40 μl/min/10(6) cells respectively). This vitro model was also used to study the metabolic interactions between BP2 and isoeugenol and coumarin. Coumarin exerted no effects on either isoeugenol or BP2 metabolism, because of its independent metabolic pathway (CYP2A6). Isoeugenol appeared to be a potent competitive substrate inhibitor of BP2 metabolism, equivalent to the specific UGT1A1 substrate: estradiol. Despite the fact that inhibition of UGT by xenobiotics is not usually considered to be a major concern, the involvement of UGT1A1 in BP2 metabolism may have pharmacokinetic and pharmacological consequences, due to the its polymorphisms in humans and its pure estrogenic effect.
Sigma-Aldrich
2,2′,4,4′-Tetrahydroxybenzophenone, 97%