Merck
  • Photoinduced Electron Transfer Switching Mechanism of a Naphthalimide Derivative with its Solvatochromic Behaviour: An Experimental and Theoretical Study with In Cell Investigations.
Photoinduced Electron Transfer Switching Mechanism of a Naphthalimide Derivative with its Solvatochromic Behaviour: An Experimental and Theoretical Study with In Cell Investigations.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-08-18)
Dipti Singharoy, Sourav Chowdhury, Soumya Sundar Mati, Swadesh Ghosh, Krishnananda Chattopadhyay, Subhash Chandra Bhattacharya
ABSTRACT
The sole existence of a t-bone-shaped naphthalimide derivative [2-(2-aminoethyl)-1H-benzo[de]isoquinoline-1,3(2H)dione] (NAP), which gives rise to a photoinduced electron transfer (PET) mechanism, has been established using a combination of experimental and theoretical studies. In parallel an in vitro-in cell PET mechanism has also been shown. To understand the photophysics of NAP, solvent studies have been carried out in different solvents. In addition, theoretical calculations have been conducted to explain the spectroscopic properties through optimized structures. A "turn off" PET mechanism has also been observed in the presence of specific metal ions, namely, Cr
Sigma-Aldrich
2-Hydroxyacetophenone, 98%