Merck
LC/MS Analysis of Methanesulfonic Acid in Busulfan Injection on SeQuant® ZIC-HILIC
LC/MS Analysis of Methanesulfonic Acid in Busulfan Injection on SeQuant® ZIC-HILIC application for LC-MS
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 3.5 μm, 200 Å (1.50444.0001)
mobile phase
[A] 100 mM ammonium acetate, pH 4.5; [B] acetonitrile; (20:80, A:B)
flow rate
0.5 mL/min
pressure
826 psi (57 bar)
column temp.
40 °C
detector
Shimadzu Prominence, RI (cell temperature  40 °C, Range 1.0, 10 μL cell)
injection
20 μL
sample
0.2 mg/ml in mobile phase
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany