Skip to Content
Merck

793025P

Avanti

DSPE-PEG2000-Tetrazine

new

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[tetrazine(polyethylene glycol)-2000] (ammonium salt)

Sign In to View Organizational & Contract Pricing

Select a Size

793025P-10MG

£281.00

£281.00


Please contact Customer Service for Availability


About This Item

Empirical Formula (Hill Notation):
C142H272N7O55P
Molecular Weight:
2988.67
UNSPSC Code:
12352211

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (793025P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NH4+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCCNC(=O)OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)Cc1ccc(cc1)-c1nncnn1)OC(=O)CCCCCCCCCCCCCCCCC

Related Categories

General description

DSPE-PEG2k-Tetrazine is a high-purity synthetic phospholipid modified with PEG2000 and a tetrazine group. PEGylation offers steric stabilization, enhancing circulation time and minimizing clearance by the reticuloendothelial system (RES). The tetrazine moiety enables bioorthogonal click chemistry, reacting selectively with trans-cyclooctene (TCO) in complex biological environments without disrupting native biological functions.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service