Skip to Content
Merck

810397P

Avanti

Sulfo-SquareFluor 631/643, free acid

new

Avanti Research - A Croda Brand

Synonym(s):

(E)-1-(5-carboxypentyl)-2-(((E)-2-hydroxy-4-oxo-3-((1,3,3-trimethyl-3H-indol-1-ium-2-yl)methylene)cyclobut-1-en-1-yl)methylene)-3,3-dimethylindoline-5-sulfonate

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C33H36N2O7S
Molecular Weight:
604.71
UNSPSC Code:
12352211

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (810397P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

fluorescence

λex 631 nm; λem 643 nm

lipid type

fluorescent lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[N+]1=C(\C=C2/C(O)=C(/C=C3/N(CCCCCC(O)=O)c4ccc(cc4C3(C)C)S([O-])(=O)=O)C2=O)C(C)(C)c2ccccc12

General description

Squaraine probes feature a distinctive squaric acid derived aromatic ring which leaves the probe vulnerable to nucleophilic attack which destroys its fluorescent ability. This limitation has historically prevented these probes from being studied as much as other probes. However, recent advancements in their synthetic design has helped address these challenges and brought them to the forefront of fluorescent imaging. Squaraines are valued for their extremely intense absorption bands, good photoconductivity, excellent photostability, and high quantum yield. 

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service