Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
Empirical Formula (Hill Notation):
C129H255N3O49
Molecular Weight:
2632.40
UNSPSC Code:
12352211
Skip To
biological source
synthetic
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 25 mg (880243P-25MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
polymerizable lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCC(=O)OCC(COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-])OC(=O)CCCCCCCCCCCCCCCCC
Related Categories
General description
DSG-PEG2000-Azide is a lipid used in drug delivery research.
Features and Benefits
The azide enables click chemistry, allowing efficient attachment of targeting ligands or other functional groups. Clickable PEG lipids support targeted delivery, and modifying the PEG length can adjust micelle size, immunogenicity, and drug release rates.
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service