Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
About This Item
Empirical Formula (Hill Notation):
C148H288N6NaO61P
Molecular Weight:
3181.84
UNSPSC Code:
12352211
Skip To
biological source
synthetic
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 5 mg (880273P-5MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
polymerizable lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Na+].C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=COOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOC=C.CCCCCCCCCCCCCCCCCC(=O)OC[C@H](
Related Categories
General description
DSPE PEG2000-DOTA free acid is a functionlized PEGylated phospholipid which can be used for LNP formulationa and other drug delivery systems.
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service