Skip to Content
Merck

880276P

Avanti

DSPE-PEG8 Biotin

new

Avanti Research - A Croda Brand

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(octaethylene glycol)] (ammonium salt)

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

Empirical Formula (Hill Notation):
C68H132N5O19PS
Molecular Weight:
1386.84
UNSPSC Code:
12352211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (880276P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

polymerizable lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[NH4+].CCCCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCCNC(=O)OCCOCCOCCOCCOCCOCCOCCOCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)OC(=O)CCCCCCCCCCCCCCCCC

General description

DSPE-PEG8 is a functionalized PEGylated phospholipid often used in drug delivery systems such as the formulation of lipid nanoparticles.

Application

Gene editing
Targeted drug and therapeutic delivery

Features and Benefits

The Biotin attachment enables site-specific attachment and the ability to create more targeted delivery systems, improving cellular uptake and therapeutic precision.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service