Skip to Content
Merck

ENA413166179

Enamine

2-(1-methyl-1H-indazol-3-yl)acetonitrile

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENA413166179 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
171.2
MDL number:
Product ENA413166179 is not currently sold in your country.

Iupac Name

(1-methyl-1H-indazol-3-yl)acetonitrile

Inchi Code

1S/C10H9N3/c1-13-10-5-3-2-4-8(10)9(12-13)6-7-11/h2-5H,6H2,1H3

InChI key

AWBGINSRBGITQS-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-140831

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-140831


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library