Skip to Content
Merck

03595

LALLS viscosity validation set polystyrene Mp 9′000-800′000 in THF certified according to DIN

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41116107

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

CofA

exact values can be taken from the accompanying certificate of analysis

Other Notes

set contains 4 different polystyrene standards with 500 mg with Mp ~9130, ~133000, ~221000, ~827000 for LALLS and viscosimeters; each standard is characterized with GPC, light scattering and viscosimetry

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service