Skip to Content
Merck

20160

n-Butyllithium solution

~1.6 M in hexanes

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
PubChem Substance ID:
UNSPSC Code:
12352103
Beilstein/REAXYS Number:
1209227
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

n-Butyllithium solution, ~1.6 M in hexanes

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

form

liquid

shelf life

limited shelf life

concentration

~1.6 M in hexanes

color

yellow

density

0.68 g/mL at 25 °C

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Disclaimer

a precipitate of lithium hydride can form on longer storage

Other Notes

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Central nervous system

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

WGK 3

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer L Rutherford et al.
Journal of the American Chemical Society, 124(2), 264-271 (2002-01-10)
6Li and (13)C NMR spectroscopic studies were carried out on [(6)Li]n-BuLi and [(6)Li]PhLi (RLi) in toluene-d(8) containing the following diamines: N,N,N',N'-tetramethylethylenediamine (TMEDA), N,N,N',N'-tetraethylethylenediamine, 1,2-dipyrrolidinoethane, 1,2-dipiperidinoethane, N,N,N',N'-tetramethylpropanediamine, trans-(R,R)-N,N,N',N'-tetramethylcyclohexanediamine, and (-)-sparteine. Dimers of general structure (RLi)(2)S(2) (S = chelating diamine) are formed
Enantioselective total synthesis of the marine toxin (-)-gymnodimine employing a Barbier-type macrocyclization.
Ke Kong et al.
Angewandte Chemie (International ed. in English), 48(40), 7402-7405 (2009-09-04)
Karl J Hale et al.
Organic letters, 5(4), 503-505 (2003-02-14)
[reaction: see text] The "Southern Hemisphere" intermediate 2, used by Masamune and co-workers for their asymmetric total synthesis of bryostatin 7 (1), has been synthesized from (E)-1,4-hexadiene (11) by a 24-step pathway that has a longest linear sequence of only
Subramanian Devaraj et al.
Carbohydrate research, 344(10), 1159-1166 (2009-06-11)
O-Alkylation of myo-inositol derivatives containing more than one hydroxyl group via their alkali metal alkoxides (sodium or lithium) preferentially occurs at a hydroxyl group having a vicinal cis-oxygen atom. In general the observed selectivity is relatively higher for lithium alkoxides
Michael A Tarselli et al.
Organic letters, 11(20), 4596-4599 (2009-10-09)
A procedure for the coupling of aliphatic imines with allylic and allenic alkoxides is described. The success of these studies was enabled by a unique reactivity profile of Ti(IV) isopropoxide/n-BuLi compared to well-known Ti(IV) isopropoxide/RMgX systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service