Merck
  • PITX1 protein interacts with ZCCHC10 to regulate hTERT mRNA transcription.
PITX1 protein interacts with ZCCHC10 to regulate hTERT mRNA transcription.
PloS one (2019-08-14)
Takahito Ohira, Hirotada Kojima, Yuko Kuroda, Sayaka Aoki, Daigo Inaoka, Mitsuhiko Osaki, Hideki Wanibuchi, Futoshi Okada, Mitsuo Oshimura, Hiroyuki Kugoh
ABSTRACT
Telomerase is a ribonucleoprotein ribonucleic enzyme that is essential for cellular immortalization via elongation of telomere repeat sequences at the end of chromosomes. Human telomerase reverse transcriptase (hTERT), the catalytic subunit of telomerase holoenzyme, is a key regulator of telomerase activity. Telomerase activity, which has been detected in the majority of cancer cells, is accompanied by hTERT expression, suggesting that this enzyme activity contributes to an unlimited replication potential of cancer cells via regulation of telomere length. Thus, hTERT is an attractive target for cancer-specific treatments. We previously reported that pared-like homeodomain 1 (PITX1) is a negative regulator of hTERT through direct binding to the hTERT promoter. However, the mechanism by which the function of PITX1 contributes to transcriptional silencing of the hTERT gene remains to be clarified. Here, we show that PITX1 and zinc finger CCHC-type containing 10 (ZCCHC10) proteins cooperate to facilitate the transcriptional regulation of the hTERT gene by functional studies via FLAG pull-down assay. Co-expression of PITX1 and ZCCHC10 resulted in inhibition of hTERT transcription, in melanoma cell lines, whereas mutate-deletion of homeodomain in PITX1 that interact with ZCCHC10 did not induce similar phenotypes. In addition, ZCCHC10 expression levels showed marked decrease in the majority of melanoma cell lines and tissues. Taken together, these results suggest that ZCCHC10-PITX1 complex is the functional unit that suppresses hTERT transcription, and may play a crucial role as a novel tumor suppressor complex.