Merck
  • Influenza A virus co-opts ERI1 exonuclease bound to histone mRNA to promote viral transcription.
Influenza A virus co-opts ERI1 exonuclease bound to histone mRNA to promote viral transcription.
Nucleic acids research (2020-09-23)
Marion Declercq, Elise Biquand, Marwah Karim, Natalia Pietrosemoli, Yves Jacob, Caroline Demeret, Cyril Barbezange, Sylvie van der Werf
ABSTRACT
Cellular exonucleases involved in the processes that regulate RNA stability and quality control have been shown to restrict or to promote the multiplication cycle of numerous RNA viruses. Influenza A viruses are major human pathogens that are responsible for seasonal epidemics, but the interplay between viral proteins and cellular exonucleases has never been specifically studied. Here, using a stringent interactomics screening strategy and an siRNA-silencing approach, we identified eight cellular factors among a set of 75 cellular proteins carrying exo(ribo)nuclease activities or involved in RNA decay processes that support influenza A virus multiplication. We show that the exoribonuclease ERI1 interacts with the PB2, PB1 and NP components of the viral ribonucleoproteins and is required for viral mRNA transcription. More specifically, we demonstrate that the protein-protein interaction is RNA dependent and that both the RNA binding and exonuclease activities of ERI1 are required to promote influenza A virus transcription. Finally, we provide evidence that during infection, the SLBP protein and histone mRNAs co-purify with vRNPs alongside ERI1, indicating that ERI1 is most probably recruited when it is present in the histone pre-mRNA processing complex in the nucleus.