Merck
  • Shedding of soluble epidermal growth factor receptor (sEGFR) is mediated by a metalloprotease/fibronectin/integrin axis and inhibited by cetuximab.
Shedding of soluble epidermal growth factor receptor (sEGFR) is mediated by a metalloprotease/fibronectin/integrin axis and inhibited by cetuximab.
Biochemistry (2013-06-05)
Jason A Wilken, Marianela Perez-Torres, Rene Nieves-Alicea, Elsa M Cora, Trace A Christensen, Andre T Baron, Nita J Maihle
ABSTRACT
Soluble epidermal growth factor receptor (sEGFR) is a circulating serum biomarker in cancer patients. Recent studies suggest that baseline serum sEGFR concentrations may predict responsiveness to EGFR-targeted therapy. Here, we demonstrate that sEGFR is generated through proteolytic cleavage of a cell surface precursor of an alternately spliced EGF receptor isoform and that sEGFR binds to EGF with high affinity. Proteolytic cleavage is stimulated by an anti-α5/β1 integrin antibody and 4-aminophenylmercuric acetate, and inhibited by fibronectin. Two FDA-approved therapeutic anti-EGFR antibodies also inhibit shedding of sEGFR, thus implicating the cell surface precursor of sEGFR as a competing target for anti-EGFR antibodies in human tissues. These observations parallel trastuzumab regulation of HER2 shedding and have implications for patient stratification in future clinical trials of EGFR-targeted antibodies.