Merck
  • LC-MS/MS quantification of next-generation biotherapeutics: a case study for an IgE binding Nanobody in cynomolgus monkey plasma.
LC-MS/MS quantification of next-generation biotherapeutics: a case study for an IgE binding Nanobody in cynomolgus monkey plasma.
Bioanalysis (2014-06-21)
Koen Sandra, Kjell Mortier, Lucie Jorge, Luis C Perez, Pat Sandra, Sofie Priem, Sofie Poelmans, Marie-Paule Bouche
ABSTRACT
Nanobodies(®) are therapeutic proteins derived from the smallest functional fragments of heavy chain-only antibodies. The development and validation of an LC-MS/MS-based method for the quantification of an IgE binding Nanobody in cynomolgus monkey plasma is presented. Nanobody quantification was performed making use of a proteotypic tryptic peptide chromatographically enriched prior to LC-MS/MS analysis. The validated LLOQ at 36 ng/ml was measured with an intra- and inter-assay precision and accuracy <20%. The required sensitivity could be obtained based on the selectivity of 2D LC combined with MS/MS. No analyte specific tools for affinity purification were used. Plasma samples originating from a PK/PD study were analyzed and compared with the results obtained with a traditional ligand-binding assay. Excellent correlations between the two techniques were obtained, and similar PK parameters were estimated. A 2D LC-MS/MS method was successfully developed and validated for the quantification of a next generation biotherapeutic.