Merck
Root-Knot and Cyst Nematodes Activate Procambium-Associated Genes in
Frontiers in plant science (2017-07-28)
Yasuka L Yamaguchi, Reira Suzuki, Javier Cabrera, Satoru Nakagami, Tomomi Sagara, Chika Ejima, Ryosuke Sano, Yuichi Aoki, Rocio Olmo, Tetsuya Kurata, Takeshi Obayashi, Taku Demura, Takashi Ishida, Carolina Escobar, Shinichiro Sawa
ABSTRACT
Developmental plasticity is one of the most striking features of plant morphogenesis, as plants are able to vary their shapes in response to environmental cues. Biotic or abiotic stimuli often promote organogenesis events in plants not observed under normal growth conditions. Root-knot nematodes (RKNs) are known to parasitize multiple species of rooting plants and to induce characteristic tissue expansion called galls or root-knots on the roots of their hosts by perturbing the plant cellular machinery. Galls contain giant cells (GCs) and neighboring cells, and the GCs are a source of nutrients for the parasitizing nematode. Highly active cell proliferation was observed in galls. However, the underlying mechanisms that regulate the symptoms triggered by the plant-nematode interaction have not yet been elucidated. In this study, we deciphered the molecular mechanism of gall formation with an